(1R,4S)-1-Methyl-4-prop-1-en-2-ylcyclohex-2-en-1-ol 95%

Lieferant: Apollo Scientific

Synonyme: (1R,4S)-1-Methyl-4-(prop-1-en-2-yl)cyclohex-2-enol

OR48128-25G OR48128-5G
APOSOR48128-25GEA 426 EUR
APOSOR48128-25G APOSOR48128-5G
(1R,4S)-1-Methyl-4-prop-1-en-2-ylcyclohex-2-en-1-ol 95%
(1R,4S)-1-Methyl-4-prop-1-en-2-ylcyclohex-2-en-1-ol
Formel: C₁₀H₁₆O
Molecular Weight: 152,233 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD27665414
CAS-Nummer: 82769-01-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR