N-Boc-N,N'-dimethylethylamine hydrochloride 97%
Lieferant: Apollo Scientific
Synonyme: N1-BOC 1,2-Dimethyl-ethylenediamine, HCl
OR48274-1G OR48274-250MG OR48274-5G
APOSOR48274-1GEA 376 EUR
APOSOR48274-1G APOSOR48274-250MG APOSOR48274-5G
N-Boc-N,N'-dimethylethylamine hydrochloride 97%
N-Boc-N,N'-dimethylethylamine hydrochloride
Formel: C₉H₂₀N₂O₂·ClH
Molecular Weight: 224.728 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13184908
CAS-Nummer: 202207-84-9