8-Mercaptoquinoline-5-sulphonic acid

Lieferant: Apollo Scientific

OR4843-10G OR4843-1G OR4843-5G
APOSOR4843-10GEA 1420 EUR
APOSOR4843-10G APOSOR4843-1G APOSOR4843-5G
8-Mercaptoquinoline-5-sulphonic acid
8-Mercaptoquinoline-5-sulphonic acid
Formel: C₉H₇NO₃S₂
Molecular Weight: 241,287 g/mol
Schmelzpunkt: 280…282 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00631699
CAS-Nummer: 5825-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR