2-Bromo-4-hydroxy-5-methoxyphenylacetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-4-(carboxymethyl)guaiacol, 5-Bromo-4-(carboxymethyl)-2-methoxyphenol

OR4895-100MG OR4895-500MG
APOSOR4895-100MGEA 209 EUR
APOSOR4895-100MG APOSOR4895-500MG
2-Bromo-4-hydroxy-5-methoxyphenylacetic acid
2-Bromo-4-hydroxy-5-methoxyphenylacetic acid
Formel: C₉H₉BrO₄
Molecular Weight: 261,069 g/mol
Schmelzpunkt: 174…177 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00079715
CAS-Nummer: 181517-98-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR