5-Chloro-2-(isopropylamino)aniline

Lieferant: Apollo Scientific
Achtung

OR4900-1G OR4900-500MG
APOSOR4900-1GEA 277 EUR
APOSOR4900-1G APOSOR4900-500MG
5-Chloro-2-(isopropylamino)aniline
5-Chloro-2-(isopropylamino)aniline
Formel: C₉H₁₃ClN₂
Molecular Weight: 184,666 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00084956
CAS-Nummer: 89659-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR