N-(2-Pyridyl)benzamide

Lieferant: Apollo Scientific

OR49047-1G OR49047-250MG
APOSOR49047-1GEA 509 EUR
APOSOR49047-1G APOSOR49047-250MG
N-(2-Pyridyl)benzamide
N-(2-Pyridyl)benzamide
Formel: C₁₂H₁₀N₂O
Molecular Weight: 198,221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00459601
CAS-Nummer: 4589-12-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR