3-(Azetidin-3-yl)propan-1-ol, N-BOC protected 95+%
Lieferant: Apollo Scientific
Synonyme: tert-Butyl 3-(3-hydroxypropyl)azetidine-1-carboxylate
OR49077-1G OR49077-250MG
APOSOR49077-1GEA 352 EUR
APOSOR49077-1G APOSOR49077-250MG
3-(Azetidin-3-yl)propan-1-ol, N-BOC protected 95+%
3-(Azetidin-3-yl)propan-1-ol, N-BOC protected
Formel: C₁₁H₂₁NO₃
Lagertemperatur: Kühlschrank
MDL: MFCD18206284
CAS-Nummer: 158602-43-8