4-Methoxycarbonyl-2,3-dihydro-1H-indole hydrochloride
Lieferant: Apollo Scientific
Synonyme: Methyl indoline-4-carboxylate hydrochloride
OR49079-1G OR49079-5G
APOSOR49079-1GEA 429 EUR
APOSOR49079-1G APOSOR49079-5G
4-Methoxycarbonyl-2,3-dihydro-1H-indole hydrochloride
4-Methoxycarbonyl-2,3-dihydro-1H-indole hydrochloride
Formel: C₁₀H₁₁NO₂·ClH
Molecular Weight: 213.661 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09026830
CAS-Nummer: 1187927-40-7