3-(2,5-Dihydro-3-methylthio-5-oxo-1,2,4-triazin-6-yl)propionic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR4911-1G
OR4911-250MG
APOSOR4911-1GEA
328
EUR
APOSOR4911-1G
APOSOR4911-250MG
3-(2,5-Dihydro-3-methylthio-5-oxo-1,2,4-triazin-6-yl)propionic acid
3-(2,5-Dihydro-3-methylthio-5-oxo-1,2,4-triazin-6-yl)propionic acid
Formel:
C₇H₉N₃O₃S Molecular Weight: 215,23 g/mol Schmelzpunkt: 228…231 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00665741 CAS-Nummer: 100524-19-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...