2-Bromo-1-(5-chloro-2-methoxyphenyl)ethanone 95+%

Lieferant: Apollo Scientific
Achtung

OR500012-100MG OR500012-250MG
APOSOR500012-100MGEA 21.9 EUR
APOSOR500012-100MG APOSOR500012-250MG
2-Bromo-1-(5-chloro-2-methoxyphenyl)ethanone 95+%
2-Bromo-1-(5-chloro-2-methoxyphenyl)ethanone
Formel: C₉H₈BrClO₂
Molecular Weight: 263,516 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01793848
CAS-Nummer: 111841-05-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR