5-Cyclopropylpicolinic acid hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR500019-100MG OR500019-250MG
APOSOR500019-100MGEA 95.7 EUR
APOSOR500019-100MG APOSOR500019-250MG
5-Cyclopropylpicolinic acid hydrochloride 97%
5-Cyclopropylpicolinic acid hydrochloride
Formel: C₉H₉NO₂·ClH
Molecular Weight: 199,634 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30730075
CAS-Nummer: 2089378-52-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR