5-Chloro-2-(methylthio)pyrimidine 97%

Lieferant: Apollo Scientific
Achtung

OR500020-1G OR500020-250MG OR500020-5G
APOSOR500020-1GEA 48 EUR
APOSOR500020-1G APOSOR500020-250MG APOSOR500020-5G
5-Chloro-2-(methylthio)pyrimidine 97%
5-Chloro-2-(methylthio)pyrimidine
Formel: C₅H₅ClN₂S
Molecular Weight: 160,625 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00489773
CAS-Nummer: 38275-42-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR