2-Bromo-4-methylthiazole-5-carbaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR500026-1G
OR500026-250MG
APOSOR500026-1GEA
180
EUR
APOSOR500026-1G
APOSOR500026-250MG
2-Bromo-4-methylthiazole-5-carbaldehyde 95%
2-Bromo-4-methylthiazole-5-carbaldehyde
Formel:
C₅H₄BrNOS Molecular Weight: 206,06 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD22041605 CAS-Nummer: 933720-87-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...