2-Bromo-5-methyl-4-nitropyridine 95%

Lieferant: Apollo Scientific
Achtung

OR50930-1G OR50930-5G OR50930-25G
APOSOR50930-1GEA 50.9 EUR
APOSOR50930-1G APOSOR50930-5G APOSOR50930-25G
2-Bromo-5-methyl-4-nitropyridine 95%
2-Bromo-5-methyl-4-nitropyridine
Formel: C₆H₅BrN₂O₂
Molecular Weight: 217,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11100652
CAS-Nummer: 66092-62-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR