4-Chloro-3-quinolinamine 95%

Lieferant: Apollo Scientific

Synonyme: 4-Chloroquinolin-3-amine

OR50943-5G
APOSOR50943-5GEA 133 EUR
APOSOR50943-5G
4-Chloro-3-quinolinamine 95%
4-Chloro-3-quinolinamine
Formel: C₉H₇ClN₂
Molecular Weight: 178,618 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00234511
CAS-Nummer: 58401-43-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR