3-Methyl-3H-benzoxazol-2-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Methylbenzo[d]oxazol-2(3H)-one
OR5097-25G
OR5097-5G
APOSOR5097-25GEA
203
EUR
APOSOR5097-25G
APOSOR5097-5G
3-Methyl-3H-benzoxazol-2-one
3-Methyl-3H-benzoxazol-2-one
Formel:
C₈H₇NO₂ Molecular Weight: 149,147 g/mol Schmelzpunkt: 82…87 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00748831 CAS-Nummer: 21892-80-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...