4-Bromo-2-phenylaniline 95%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-[1,1'-biphenyl]-2-amine

OR50982-1G OR50982-250MG OR50982-5G
APOSOR50982-1GEA 478 EUR
APOSOR50982-1G APOSOR50982-250MG APOSOR50982-5G
4-Bromo-2-phenylaniline 95%
4-Bromo-2-phenylaniline
Formel: C₁₂H₁₀BrN
Molecular Weight: 248,119 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09754386
CAS-Nummer: 5455-13-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR