2-Bromo-6-methoxybenzonitrile 98%

Lieferant: Apollo Scientific

OR50983-1G OR50983-25G OR50983-5G
APOSOR50983-1GEA 31.9 EUR
APOSOR50983-1G APOSOR50983-25G APOSOR50983-5G
2-Bromo-6-methoxybenzonitrile 98%
2-Bromo-6-methoxybenzonitrile
Formel: C₈H₆BrNO
Molecular Weight: 212,043 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09834771
CAS-Nummer: 1245647-50-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR