3-Bromo-2-nitrophenylacetic acid

Lieferant: Apollo Scientific
Achtung

OR50991-1G OR50991-250MG
APOSOR50991-1GEA 719 EUR
APOSOR50991-1G APOSOR50991-250MG
3-Bromo-2-nitrophenylacetic acid
3-Bromo-2-nitrophenylacetic acid
Formel: C₈H₆BrNO₄
Molecular Weight: 260.042 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18392318
CAS-Nummer: 400629-31-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR