3-Bromo-4-nitrophenylacetic acid

Lieferant: Apollo Scientific
Achtung

OR50992-1G OR50992-250MG OR50992-5G
APOSOR50992-1GEA 261 EUR
APOSOR50992-1G APOSOR50992-250MG APOSOR50992-5G
3-Bromo-4-nitrophenylacetic acid
3-Bromo-4-nitrophenylacetic acid
Formel: C₈H₆BrNO₄
Molecular Weight: 260,042 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11848704
CAS-Nummer: 90004-95-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR