(1R)-2-Chloro-1-(4-chlorophenyl)ethan-1-ol

Lieferant: Apollo Scientific

OR51000-100MG OR51000-1G OR51000-250MG
APOSOR51000-100MGEA 58 EUR
APOSOR51000-100MG APOSOR51000-1G APOSOR51000-250MG
(1R)-2-Chloro-1-(4-chlorophenyl)ethan-1-ol
(1R)-2-Chloro-1-(4-chlorophenyl)ethan-1-ol
Formel: C₈H₈Cl₂O
Molecular Weight: 191.055 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09863568
CAS-Nummer: 178460-80-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR