Ethyl 2-amino-5-bromo-1H-indole-3-carboxylate ≥97%

Lieferant: Apollo Scientific

OR510114-250MG
APOSOR510114-250MGEA 65.4 EUR
APOSOR510114-250MG
Ethyl 2-amino-5-bromo-1H-indole-3-carboxylate ≥97%
Ethyl 2-amino-5-bromo-1H-indole-3-carboxylate
Formel: C₁₁H₁₁BrN₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD20226197
CAS-Nummer: 1242140-62-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR