(9,9-Diphenylfluoren-4-yl)boronic acid 99%
Lieferant: Apollo Scientific
Synonyme:
(9,9-Diphenyl-9H-fluoren-4-yl)boronic acid
OR51018-5G
APOSOR51018-5GEA
65.4
EUR
APOSOR51018-5G
(9,9-Diphenylfluoren-4-yl)boronic acid 99%
(9,9-Diphenylfluoren-4-yl)boronic acid
Formel:
C₂₅H₁₉BO₂ Molecular Weight: 362,228 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29077672 CAS-Nummer: 1224976-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...