2-Bromo-7-chlorospiro[fluorene-9,9'-xanthene] 99%

Lieferant: Apollo Scientific

OR51029-1G OR51029-5G OR51029-10G
APOSOR51029-1GEA 139 EUR
APOSOR51029-1G APOSOR51029-5G APOSOR51029-10G
2-Bromo-7-chlorospiro[fluorene-9,9'-xanthene] 99%
2-Bromo-7-chlorospiro[fluorene-9,9'-xanthene]
Formel: C₂₅H₁₄BrClO
Molecular Weight: 445,735 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32708890
CAS-Nummer: 2259716-83-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR