7-Bromo-2-iodo-1-(phenylsulphonyl)-1H-indole 95%
Lieferant: Apollo Scientific
Synonyme:
1-(Benzenesulphonyl)-7-bromo-2-iodo-1H-indole
OR51177-1G
OR51177-250MG
APOSOR51177-1GEA
654
EUR
APOSOR51177-1G
APOSOR51177-250MG
7-Bromo-2-iodo-1-(phenylsulphonyl)-1H-indole 95%
7-Bromo-2-iodo-1-(phenylsulphonyl)-1H-indole
Formel:
C₁₄H₉BrINO₂S Molecular Weight: 462.1 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
2514953-08-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...