2-(Methoxymethyl)benzaldehyde 95%

Lieferant: Apollo Scientific

OR51189-1G OR51189-250MG
APOSOR51189-1GEA 1200 EUR
APOSOR51189-1G APOSOR51189-250MG
2-(Methoxymethyl)benzaldehyde 95%
2-(Methoxymethyl)benzaldehyde
Formel: C₉H₁₀O₂
Molecular Weight: 150.175 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD24395542
CAS-Nummer: 106020-70-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR