2-Cyano-3-hydroxybenzoic acid 95%

Lieferant: Apollo Scientific

OR51344-1G OR51344-250MG
APOSOR51344-1GEA 1090 EUR
APOSOR51344-1G APOSOR51344-250MG
2-Cyano-3-hydroxybenzoic acid 95%
2-Cyano-3-hydroxybenzoic acid
Formel: C₈H₅NO₃
Molecular Weight: 163,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17000076
CAS-Nummer: 1243282-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR