tert-Butyl N-amino-N-[(2-bromophenyl)methyl]carbamate 95%
Lieferant: Apollo Scientific
OR51363-1G
OR51363-250MG
APOSOR51363-1GEA
609
EUR
APOSOR51363-1G
APOSOR51363-250MG
tert-Butyl N-amino-N-[(2-bromophenyl)methyl]carbamate 95%
tert-Butyl N-amino-N-[(2-bromophenyl)methyl]carbamate
Formel:
C₁₂H₁₇BrN₂O₂ Molecular Weight: 301,18 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
2268038-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...