5-Bromo-4-methyl-3-nitro-1H-pyrazole 95%
Lieferant: Apollo Scientific
OR51557-1G
OR51557-250MG
APOSOR51557-1GEA
719
EUR
APOSOR51557-1G
APOSOR51557-250MG
5-Bromo-4-methyl-3-nitro-1H-pyrazole 95%
5-Bromo-4-methyl-3-nitro-1H-pyrazole
Formel:
C₄H₄BrN₃O₂ Molecular Weight: 205.997 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32689343 CAS-Nummer: 2514952-92-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...