5-Bromo-4-methyl-1H-pyrazol-3-amine 95%

Lieferant: Apollo Scientific

OR51558-1G OR51558-250MG
APOSOR51558-1GEA 799 EUR
APOSOR51558-1G APOSOR51558-250MG
5-Bromo-4-methyl-1H-pyrazol-3-amine 95%
5-Bromo-4-methyl-1H-pyrazol-3-amine
Formel: C₄H₆BrN₃
Molecular Weight: 176.015 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32689344
CAS-Nummer: 2514952-93-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR