4-Acetamido-3-nitrobenzonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR51724-1G
OR51724-5G
APOSOR51724-1GEA
0
EUR
APOSOR51724-1G
APOSOR51724-5G
4-Acetamido-3-nitrobenzonitrile
4-Acetamido-3-nitrobenzonitrile
Formel:
C₉H₇N₃O₃ Schmelzpunkt: 124…127 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00724165 CAS-Nummer: 29289-18-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...