5-Bromo-3-methylpyridine-2-carboxaldehyde 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-3-methylpicolinaldehyde, 5-Bromo-2-formyl-3-methylpyridine, 5-Bromo-3-methyl-2-pyridinecarbaldehyde
OR51763-1G
OR51763-250MG
APOSOR51763-1GEA
150
EUR
APOSOR51763-1G
APOSOR51763-250MG
5-Bromo-3-methylpyridine-2-carboxaldehyde 97%
5-Bromo-3-methylpyridine-2-carboxaldehyde
Formel:
C₇H₆BrNO Molecular Weight: 200,033 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD07776420 CAS-Nummer: 376587-53-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...