2-Bromo-5-chloro-4-hydroxypyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-5-chloropyridin-4-ol

OR51774-1G OR51774-250MG
APOSOR51774-1GEA 281 EUR
APOSOR51774-1G APOSOR51774-250MG
2-Bromo-5-chloro-4-hydroxypyridine
2-Bromo-5-chloro-4-hydroxypyridine
Formel: C₅H₃BrClNO
Molecular Weight: 208,44 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13189879
CAS-Nummer: 1196145-66-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR