2,3-Dichloro-4-hydroxypyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,3-Dichloropyridin-4-ol
OR51779-1G
OR51779-5G
APOSOR51779-1GEA
263
EUR
APOSOR51779-1G
APOSOR51779-5G
2,3-Dichloro-4-hydroxypyridine
2,3-Dichloro-4-hydroxypyridine
Formel:
C₅H₃Cl₂NO Molecular Weight: 163,989 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16610774 CAS-Nummer: 1174047-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...