5-Hydroxy-N-methylpyridine-2-carboxamide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Hydroxy-N-methylpicolinamide, 5-Hydroxy-2-(methylcarbamoyl)pyridine
OR51785-1G
OR51785-250MG
APOSOR51785-1GEA
326
EUR
APOSOR51785-1G
APOSOR51785-250MG
5-Hydroxy-N-methylpyridine-2-carboxamide
5-Hydroxy-N-methylpyridine-2-carboxamide
Formel:
C₇H₈N₂O₂ Molecular Weight: 152.151 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11977447 CAS-Nummer: 859538-76-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...