(2R,6R)-2,6-Dimethylmorphline 97%

Lieferant: Apollo Scientific

Synonyme: (2R,6R)-2,6-Dimethylmorpholine

OR51787-250MG OR51787-1G
APOSOR51787-250MGEA 277 EUR
APOSOR51787-250MG APOSOR51787-1G
(2R,6R)-2,6-Dimethylmorphline 97%
(2R,6R)-2,6-Dimethylmorphline
Formel: C₆H₁₃NO
Molecular Weight: 115,174 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18250270
CAS-Nummer: 171753-74-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR