6-Chloro-5-iodo-2-(methylsulphonyl)-1H-imidazo[4,5-b]pyridine 95%

Lieferant: Apollo Scientific
Achtung

OR51796-100MG OR51796-250MG
APOSOR51796-100MGEA 58 EUR
APOSOR51796-100MG APOSOR51796-250MG
6-Chloro-5-iodo-2-(methylsulphonyl)-1H-imidazo[4,5-b]pyridine 95%
6-Chloro-5-iodo-2-(methylsulphonyl)-1H-imidazo[4,5-b]pyridine
Formel: C₇H₅ClIN₃O₂S
Molecular Weight: 357,556 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28965894
CAS-Nummer: 1394373-18-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR