2-Bromo-6-cyclopropylphenol

Lieferant: Apollo Scientific
Achtung

OR51799-1G OR51799-250MG OR51799-5G
APOSOR51799-1GEA 114 EUR
APOSOR51799-1G APOSOR51799-250MG APOSOR51799-5G
2-Bromo-6-cyclopropylphenol
2-Bromo-6-cyclopropylphenol
Formel: C₉H₉BrO
Molecular Weight: 213,071 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD24719347
CAS-Nummer: 911817-98-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR