2-Iodo-6-methylbenzyl alcohol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Iodo-6-methylphenyl)methanol, (6-Iodo-o-tolyl)methanol

OR51813-1G OR51813-250MG OR51813-5G
APOSOR51813-1GEA 288 EUR
APOSOR51813-1G APOSOR51813-250MG APOSOR51813-5G
2-Iodo-6-methylbenzyl alcohol 95%
2-Iodo-6-methylbenzyl alcohol
Formel: C₈H₉IO
Molecular Weight: 248,061 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18397812
CAS-Nummer: 1261794-19-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR