2-Iodo-6-methylbenzaldehyde 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Formyl-3-iodotoluene, 6-Iodo-o-tolualdehyde

OR51863-1G OR51863-250MG OR51863-5G
APOSOR51863-1GEA 288 EUR
APOSOR51863-1G APOSOR51863-250MG APOSOR51863-5G
2-Iodo-6-methylbenzaldehyde 95%
2-Iodo-6-methylbenzaldehyde
Formel: C₈H₇IO
Molecular Weight: 246,045 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18397773
CAS-Nummer: 1261826-51-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR