2,2-Dimethoxybutane 90%

Lieferant: Apollo Scientific
Achtung

OR51925-1G OR51925-5G
APOSOR51925-1GEA 184 EUR
APOSOR51925-1G APOSOR51925-5G
2,2-Dimethoxybutane 90%
2,2-Dimethoxybutane
Formel: C₆H₁₄O₂
Molecular Weight: 118.174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185945
CAS-Nummer: 3453-99-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR