3-(Aminomethyl)-5-methyl-2-phenylfuran 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: (5-Methyl-2-phenylfur-3-yl)methylamine

OR5207-1G OR5207-250MG
APOSOR5207-1GEA 524 EUR
APOSOR5207-1G APOSOR5207-250MG
3-(Aminomethyl)-5-methyl-2-phenylfuran 97%
3-(Aminomethyl)-5-methyl-2-phenylfuran
Formel: C₁₂H₁₃NO
Molecular Weight: 187,238 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06213531
CAS-Nummer: 771572-29-3
UN: 2922
ADR: 8+6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR