Methyl 3,5-dichloro-2-hydroxybenzoate 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,4-Dichloro-6-(methoxycarbonyl)phenol, Methyl 3,5-dichlorosalicylate
OR52077-1G
OR52077-5G
OR52077-250MG
APOSOR52077-1GEA
256
EUR
APOSOR52077-1G
APOSOR52077-5G
APOSOR52077-250MG
Methyl 3,5-dichloro-2-hydroxybenzoate 97%
Methyl 3,5-dichloro-2-hydroxybenzoate
Formel:
C₈H₆Cl₂O₃ Molecular Weight: 221,037 g/mol Schmelzpunkt: 142…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00464939 CAS-Nummer: 7606-87-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...