4-Bromo-2-chloro-1-methyl-1H-benzimidazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-2-chloro-1-methyl-1H-benzo[d]imidazole
OR52093-100MG
OR52093-500MG
OR52093-1G
APOSOR52093-100MGEA
145
EUR
APOSOR52093-100MG
APOSOR52093-500MG
APOSOR52093-1G
4-Bromo-2-chloro-1-methyl-1H-benzimidazole
4-Bromo-2-chloro-1-methyl-1H-benzimidazole
Formel:
C₈H₆BrClN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD17168994 CAS-Nummer: 1240610-45-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...