6-Bromo-2-chloro-1-methyl-1H-benzimidazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Bromo-2-chloro-1-methyl-1H-benzo[d]imidazole
OR52118-100MG
OR52118-500MG
APOSOR52118-100MGEA
0
EUR
APOSOR52118-100MG
APOSOR52118-500MG
6-Bromo-2-chloro-1-methyl-1H-benzimidazole
6-Bromo-2-chloro-1-methyl-1H-benzimidazole
Formel:
C₈H₆BrClN₂ Molecular Weight: 245,504 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18073561 CAS-Nummer: 1378947-22-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...