N-Methyl-2-(4-methylhomopiperazin-1-yl)benzylamine 97%

Lieferant: Apollo Scientific

Synonyme: N-Methyl-2-(4-methylperhydro-1,4-diazepin-1-yl)benzylamine

OR5213-1G OR5213-250MG
APOSOR5213-1GEA 230 EUR
APOSOR5213-1G APOSOR5213-250MG
N-Methyl-2-(4-methylhomopiperazin-1-yl)benzylamine 97%
N-Methyl-2-(4-methylhomopiperazin-1-yl)benzylamine
Formel: C₁₄H₂₃N₃
Molecular Weight: 233,353 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09065003
CAS-Nummer: 915707-57-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR