4-(4-Methylhomopiperazin-1-yl)aniline trihydrochloride hydrate

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(4-Methyl-1,4-diazepan-1-yl)aniline trihydrochloride hydrate

OR5215-1G OR5215-250MG
APOSOR5215-1GEA 256 EUR
APOSOR5215-1G APOSOR5215-250MG
4-(4-Methylhomopiperazin-1-yl)aniline trihydrochloride hydrate
4-(4-Methylhomopiperazin-1-yl)aniline trihydrochloride hydrate
Formel: C₁₂H₁₉N₃·3ClH·H₂O
Molecular Weight: 332,697 g/mol
Schmelzpunkt: 238…250 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09026608
CAS-Nummer: 913830-33-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR