4-Allyloxy2,2,6,6-tetramethylpiperidine 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,2,6,6-Tetramethyl-4-(prop-2-en-1-yloxy)piperidine
OR52153-1G
OR52153-5G
APOSOR52153-1GEA
168
EUR
APOSOR52153-1G
APOSOR52153-5G
4-Allyloxy2,2,6,6-tetramethylpiperidine 95%
4-Allyloxy2,2,6,6-tetramethylpiperidine
Formel:
C₁₂H₂₃NO Molecular Weight: 197,317 g/mol Dichte: 0,88…0.92 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD23381681 CAS-Nummer: 43224-75-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...