[2-(Benzyloxy)ethoxy]acetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: BnO-PEG1-CH2COOH, 2-(2-(Benzyloxy)ethoxy)acetic acid

OR52176-1G OR52176-250MG
APOSOR52176-1GEA 151 EUR
APOSOR52176-1G APOSOR52176-250MG
[2-(Benzyloxy)ethoxy]acetic acid
[2-(Benzyloxy)ethoxy]acetic acid
Formel: C₁₁H₁₄O₄
Molecular Weight: 210,226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16294180
CAS-Nummer: 93206-09-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR