3-Amino-5-bromo-2-methoxybenzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-3-carboxy-2-methoxyaniline
OR52230-1G
OR52230-250MG
OR52230-5G
APOSOR52230-1GEA
986
EUR
APOSOR52230-1G
APOSOR52230-250MG
APOSOR52230-5G
3-Amino-5-bromo-2-methoxybenzoic acid
3-Amino-5-bromo-2-methoxybenzoic acid
Formel:
C₈H₈BrNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD28384162 CAS-Nummer: 60541-85-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...